missing translation for 'onlineSavingsMsg'
Learn More

Sodium hydroxide concentrate, For 1 L standard solution, 1.0 M NaOH (1.0 N), Honeywell Fluka™

for 1 L standard solution, 1.0 M NaOH (1.0 N)

£48.15 - £228.96

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15635250
View Documents Promotion Details
Honeywell-Fluka
38215-1EA
1 L
£48.15
Each
Please to purchase this item. Need a web account? Register with us today!
15645250
View Documents Promotion Details
Honeywell Fluka
38215-6X1EA
6 x 1 Ea.
£228.96
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]

Specifications

1310-73-2
NaOH
UN1824
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40g/mol
3 mmHg (37°C)
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Ampoule
Sodium hydroxide concentrate
Videos
Safety and Handling

Safety and Handling

P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338

Product Identifier
  • Sodium hydroxide concentrate

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide; caustic soda

SDS
Documents

Documents

Product Certifications