missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide Concentrate, for 1M solution, Honeywell Fluka™

Product Code. 15695240
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 2L standard solution, 1 M NaOH (1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1310-73-2
Molecular Formula HNaO
Linear Formula NaOH
MDL Number MFCD00003548
UN Number UN1824
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 Pc.
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40g/mol
Vapor Pressure 3 mmHg (37°C)
Chemical Name or Material Sodium hydroxide concentrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'