missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide, BAKER, J.T. Baker™
64.22 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
Description
A grade of chemicals for laboratory use, for organic synthesis, and for routine research usually controlled by physical criteria and often by assay (by GC).
Specifications
Color | White |
Boiling Point | 1390°C |
Melting Point | 319°C |
Assay Percent Range | 97.0–100.5% |
Density | 2.13g/cm3 at 20°C |
Solubility Information | In water: 420g/L at 20°C |
Quantity | 1 kg |
Formula Weight | 39.997g/mol |
Grade | Meets reagent specifications for testing Ph. Eur. monographs |
Identification | Passes test |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'