missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydroxide, 2.5M, , Analytical Volumetric Solution, Reagecon™
BIOPHARMA

Product Code. 13146309
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in analytical chemistry as a titration solution.

  • Molecular Formula: NaOH
  • Molar Mass: 40.00g/mol
  • CAS: 1310-73-2
  • EIN ECS No.: 215-185-5

Specifications

Color Colorless
Boiling Point 100°C
CAS 1310-73-2
Molecular Formula HNaO
Concentration 2.5mol/L
Density 1.10433g/cm3
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Fully miscible
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Quantity 1 L
SMILES [OH-].[Na+]
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
PubChem CID 14798
ChEBI CHEBI:32145
Formula Weight 40.00g/mol
Normality 2.5N
Vapor Pressure 23hPa (at 20°C)
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • SODIUM HYDROXIDE 2.5N (2.5M)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'