Learn More
Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7775-14-6 |
---|---|
MDL Number | MFCD00011640 |
InChI Key | JVBXVOWTABLYPX-UHFFFAOYSA-L |
Synonym | sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros |
PubChem CID | 24489 |
ChEBI | CHEBI:66870 |
SMILES | [O-]S(=O)S(=O)[O-].[Na+].[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
7775-14-6 | |
JVBXVOWTABLYPX-UHFFFAOYSA-L | |
24489 | |
[O-]S(=O)S(=O)[O-].[Na+].[Na+] |
MFCD00011640 | |
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
CHEBI:66870 |
Specifications
>300.0°C | |
White | |
ca. 85% | |
Na2S2O4 | |
15, 8747 | |
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
[O-]S(=O)S(=O)[O-].[Na+].[Na+] | |
24489 | |
174.1 | |
Technical |
7775-14-6 | |
Crystalline Powder | |
Na2O4S2 | |
MFCD00011640 | |
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
JVBXVOWTABLYPX-UHFFFAOYSA-L | |
174.1 | |
CHEBI:66870 | |
∼85% | |
Sodium hydrosulfite, tech., ca. 85% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Self-heating; may catch fire.
Contact with acids liberates toxic gas.
GHS P Statement
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-890-0
missing translation for 'tsca' : TSCA
- Sodium hydrosulfite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Self-heating substance Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H251-Self-heating: may catch fire.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P235+P410-Keep cool. Protect from sunlight.
- P264-Wash thoroughly after handling.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only