missing translation for 'onlineSavingsMsg'
Learn More

Sodium Hydrosulfide Hydrate, Honeywell Fluka™

Product Code. 15681530
Click to view available options
Quantity:
5 g
100 g
1 kg
3 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

min. 60.0% (NaHS)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207683-19-0
Molecular Formula HNaS
Molecular Weight (g/mol) 56.06
MDL Number MFCD00149796
InChI Key HYHCSLBZRBJJCH-UHFFFAOYSA-M
Synonym sodium hydrosulfide hydrate, hnas.h2o, nahs.h2o, sodium sulfanide hydrate, sodium bisulfide monohydrate, sodium hydrosulphide hydrate, sodium hydro sulfide hydrate, sodium hydrosulfide monohydrate, sodium hydrate sulfanide, sodium hydrosulfide hydrate, cp
PubChem CID 16211346
SMILES [Na+].[SH-]

Specifications

Melting Point 52°C to 54°C
Color Yellow
Quantity 100 g
Linear Formula NaSH·xH2O
UN Number UN2949
Flash Point 90°C (194°F)
IUPAC Name sodium;sulfanide;hydrate
Formula Weight 56.06g/mol (anhydrous basis)
Packaging Glass Bottle
Appearance Flakes
Chemical Name or Material Sodium Hydrosulfide Hydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'