missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium citrate, 0.5M buffer soln., pH 6.0

£61.71 - £91.35

Specifications

Chemical Name or Material Sodium citrate buffer soln.
Color Colorless
Concentration 0.5 M
pH 6.0
Physical Form Liquid
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15494319
View Documents Promotion Details
Thermo Scientific Chemicals
J61815.AK
250 mL
£61.71
250mL
Please to purchase this item. Need a web account? Register with us today!
15404329
View Documents Promotion Details
Thermo Scientific Chemicals
J61815.AP
500 mL
£91.35
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium citrate, 0.5M buffer soln., pH 6.0 is used for RNA isolation. It aids in the detection of antigens in fixed tissue preparations.

Solubility
Fully miscible in water.

Notes
Incompatible with heat and oxidizing agents.
Specifications

Specifications

Sodium citrate buffer soln.
0.5 M
Liquid
MFCD00012462
Colorless
6.0
Odorless
Fully miscible in water.
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only