missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Honeywell Fluka™
Click to view available options
Quantity:
1 kg
2.5 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 6 x 1 kg |
Linear Formula | Na2CO3 · 10H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 286.14 |
Percent Purity | 99.5 to 100.5% (ex dried subst.) |
Grade | puriss. |
Chemical Name or Material | Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'