missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bromide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
IUPAC Name | sodium bromide |
Formula Weight | 102.89 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 3.21kg |
Chemical Name or Material | Sodium Bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'