missing translation for 'onlineSavingsMsg'
Learn More

Sodium Bromide, ≥99%, ReagentPlus™, Honeywell Fluka™

Reagent Grade,≥99%

£27.83 - £507.92

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
SMILES [Na+].[Br-]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15602080
View Documents Promotion Details
Honeywell Research Chemicals
220345-500G
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15692070
View Documents Promotion Details
Honeywell Research Chemicals
220345-2.5KG
2.5 kg Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15612080
View Documents Promotion Details
Honeywell Research Chemicals
220345-6X500G
6 x 500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15682070
View Documents Promotion Details
Honeywell Research Chemicals
220345-12KG
12 kg Drum with inliner
£507.92
12kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

7647-15-6
102.89
JHJLBTNAGRQEKS-UHFFFAOYSA-M
253881
[Na+].[Br-]
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
CHEBI:63004

Specifications

7647-15-6
NaBr
NONH for all modes of transport
Soluble in ethanol, Soluble in water
[Na+].[Br-]
102.89
CHEBI:63004
≥99%
1mmHg (806°C)
BrNa
MFCD00003475
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
JHJLBTNAGRQEKS-UHFFFAOYSA-M
sodium bromide
253881
102.89g/mol
ReagentPlus™
Sodium Bromide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-599-9

missing translation for 'rtecsNumber' : VZ3150000

SDS
Documents

Documents

Product Certifications