Learn More
SOD2 Mouse anti-Human, CoraLite 488, Clone: 3A6C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66474-100UL
Additional Details : Weight : 0.00010kg
Description
The SOD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOD2. This antibody recognizes human, mouse, rat, pig antigen. The SOD2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specifications
SOD2 | |
Monoclonal | |
CoraLite 488 | |
100 μL | |
6648 | |
IgG2a |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
3A6C2 | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |