Learn More
Thermo Scientific™ SLIPFREE Ferrules
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Brand: Thermo Scientific™ 36023
Code : 50
Additional Details : Weight : 0.10000kg
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
- Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
- Kel-F and PEEK are offered for applications in which Vespel is not suitable
Specifications
SLIPFREE Connectors for HPLC Columns | |
PEEK sealing ferrule | |
PEEK | |
Sealing Ferrule |