Learn More
SLC22A12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555113
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ESARWLLTTG RLDWGLQELW RVAAINGKGA VQDTLTPEVL LSAMREELSM GQPPASLGTL LRMPGLRFRT CI Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 64%.
SLC22A12 is required for efficient urate re-absorption in the kidney. SLC22A12 regulates blood urate levels. SLC22A12 mediates saturable urate uptake by facilitating the exchange of urate against organic anions.The protein encoded by this gene is a urate transporter and urate-anion exchanger which regulates the level of urate in the blood. This protein is an integral membrane protein primarily found in kidney. Two transcript variants encoding different isoforms have been found for this gene.Specifications
SLC22A12 | |
Polyclonal | |
Unconjugated | |
SLC22A12 | |
AI987855; OAT4L; OATL4; Organic anion transporter 4-like protein; renal-specific transporter; RST; Slc22a12; Slc22al2; solute carrier family 22 (organic anion/cation transporter), member 12; solute carrier family 22 (organic anion/urate transporter), member 12; solute carrier family 22 (organic cation transporter)-like 2; solute carrier family 22 member 12; UNQ6453/PRO34004; URAT1; Urate anion exchanger 1; urate transporter 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116085 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96S37 | |
SLC22A12 | |
Recombinant protein corresponding to Human SLC22A12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only