missing translation for 'onlineSavingsMsg'
Learn More

CD96 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met 1-Met 503) fused to a polyhistidine tag at the C-terminus

£377.00 - £1204.00

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias CD96
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15258365
View Documents
Invitrogen
11202H08H50
50μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15238365
View Documents
Invitrogen
11202H08H250
5 x 50μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Cluster of Differentiation 96 (CD96), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: 1. Measured by its ability to bind recombinant mouse PVR in a functional ELISA; 2. Measured by its ability to bind recombinant Human CD155 in a functional ELISA
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥85% by SDS-PAGE
<1 EU/μg
CD96
PAGE
Lyophilized
10225

For Research Use Only. Not for use in diagnostic procedures.