Learn More
Simport™ Scientific Sample Tubes
For transport and storage of urine, sputum and most liquids or particulate samples
Description
- Tube made of polypropylene
- Cap made of polyethylene
- Conical bottom tubes with self-standing base graduated
- Chemically resistant and shatterproof
- Supplied with a leakproof screw cap
- Molded-in graduations make them easy to read
For transporting liquid sample/hazardous material