missing translation for 'onlineSavingsMsg'
Learn More

Silver acetate, 99%, pure, Thermo Scientific Chemicals

Catalog Number p-221885
Click to view available options
Quantity:
25 g
100 g
500 g
563-63-3
C2H3AgO2
166.91
MFCD00012446
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
11246
silver;acetate
CC(=O)[O-].[Ag+]
563-63-3
C2H3AgO2
166.91
MFCD00012446
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
11246
silver;acetate
CC(=O)[O-].[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY
Color Beige-Gray to White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula CH3CO2Ag
Fieser 01,1002; 03,362; 06,511; 08,440; 12,256; 13,285
Merck Index 15,8644
Solubility Information Solubility in water: 10.2g/L (20°C). Other solubilities: soluble in nitric acid
Formula Weight 166.91
Percent Purity 99%
Grade Pure
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 3.25
Density 3.2500g/mL
Chemical Name or Material Silver acetate, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'