missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heidolph instruments Silicone Tubing
Platinum-coated silicone tubing for use in pharmaceuticals and biology. Heidolph™ Silicone Tubing is very smooth, biocompatible and inert.
£29.00 - £55.00
Product Code | Brand | Tubing Size | Thickness (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Thickness (Metric) | Price | Quantity & Availability | ||||
10721103
|
Heidolph instruments
525-34000-00 |
0.5 | 1.6 mm |
£31.00
1m |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
10361582
|
Heidolph instruments
525-30027-00 |
4.8 | 2.5 mm |
£36.00
1m |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
10660303
|
Heidolph instruments
525-30028-00 |
6.4 | 1.6 mm |
£29.00
1m |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
10585112
|
Heidolph instruments
525-39000-00 |
6.4 | 2.5 mm |
£55.00
1m |
Estimated Shipment: 30-09-2024
Log in to see stock. |
||||
Description
- Extremely smooth interior prevents bacterial growth.
- Biocompatible, minimal adsorption and absorption.
- Good flow properties.
- High temperature stability.
- Absolutely inert, softener-free.
Specifications
Autoclavable | |
Non-sterile |