missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ SILBERBRAND Compact Burette Spare Amber Burette Tube
SILBERBRAND Spare Burette Tube is suitable for use with a compact burette.
£49.70 - £50.20
Specifications
Color | Amber |
---|---|
For Use With (Equipment) | Compact Burette |
Material | Borosilicate Glass (5.4) |
Description
- Constructed of Borosilicate Glass 3.3
- Amber glass
Specifications
Amber | |
Compact Burette | |
Borosilicate Glass (5.4) |