missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SHISA9 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1172
Additional Details : Weight : 0.01000kg
Description
SHISA9 Synthetic Peptide, PEP-1172, from Invitrogen.
This peptide corresponds to 22 amino acids near the center of human SHISA9.PEP-1172 can be used as a blocking peptide with polyclonal antibody PA5-21058.Specifications
729993 | |
purified | |
Human SHISA9 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
B4DS77 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
SHISA9 | |
0.02% sodium azide | |
50 μg | |
PBS |