Learn More
Epredia™ Shandon™ Colormark™ Slides, white, bulk
Use Epredia™ Shandon™ Colormark™ Slides with the Epredia™ Shandon™ MicroWriter™ Slide Labeler.
Brand: Epredia™ B9992000
Code : NEW
Additional Details : Weight : 0.64800kg
Specifications
Microscope Slide | |
Glass | |
White | |
144/Pk. |