missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D3A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | SH2D3A |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16814904
|
Proteintech
15641-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16804904
|
Proteintech
15641-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play a role in JNK activation.Specifications
SH2D3A | |
Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9BRG2 | |
10045 | |
SH2D3A Fusion Protein Ag8132 | |
Primary | |
-20°C | |
SH2D3A |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Novel SH2 containing protein 1, NSP1, SH2 domain containing 3A, SH2D3A | |
SH2D3A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |