Learn More
Trajan™ Thermo Scientific Instrument Ferrules
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand: Trajan™ 0726549
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Next generation ferrule system for gas chromatography
- Eliminates leaks
- No contamination
- No need to retighten after temperature cycling
- Cleaner spectra for better library matches
Specifications
Graphite, Vespel | |
Detector/Injectors | |
0.10 to 0.25 mm |