missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Stylet Wires for Syringe Cleaning

Designed for effective cleaning of syringe needles. Trajan Scientific and Medical™ Stylet Wires for Syringe Cleaning are supplied in packs of 5.

Brand:  Trajan™ 031745

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 15258523

  • £14.05 / Pack of 5
Estimated Shipment: 10-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Designed for effective cleaning of syringe needles. Trajan Scientific and Medical™ Stylet Wires for Syringe Cleaning are supplied in packs of 5.

  • Effective cleaning of syringe needle
  • Kit available as well as replacement wires
Specifications

Specifications

5.9 in.
15 cm
Replacement Stylet Wire
Syringe Needles (5 to 10μL)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers