Learn More
Trajan™ Gas and Vapor Sampler Bag
Enables spot collection of environmental gas samples or low pressure gas streams in 200cm3 impervious resealable bags
Brand: Trajan™ 090112
Additional Details : Weight : 2.00000kg
Description
Specifications
Sample Bag | |
Ideal for field analysis | |
Impervious | |
10/Pk. |