Learn More
SGE™ BP20 (WAX) Capillary Columns
Industry standard WAX columns
£530.00 - £1025.00
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
15278673
|
SGE™
054439 |
0.25 mm | 1 μm | 30 m |
£530.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
12935591
|
SGE™
054438 |
0.32 mm | 0.5 μm | 30 m |
£565.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
12664955
|
SGE™
054427 |
0.25 mm | 0.25 μm | 30 m |
£555.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
12965581
|
SGE™
054433 |
0.32 mm | 0.25 μm | 30 m |
£565.00
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
12995581
|
SGE™
054451 |
0.53 mm | 1 μm | 30 m |
£620.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
15248683
|
SGE™
0544515 |
0.53 mm | 1 μm | 60 m |
£1025.00
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
Description
- Very polar polyethylene glycol (PEG) phase
- Cross-linked for stability and washing
- Also ideal for xylene and aromatic isomer
- 0.1 to 1μm film thickness: 20° to 260/280°C
Specifications
SGE™ BP20 (Wax) | |
Polar |