Learn More
Invitrogen™ Serpin A4 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHSERPINA4X5
Additional Details : Weight : 0.33820kg
Description
The Human Serpin A4 ELISA quantitates Hu Serpin A4 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Serpin A4. Principle of the method The Human Serpin A4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Inhibits human amidolytic and kininogenase activities of tissue kallikrein. Inhibition is achieved by formation of an equimolar, heat- and SDS-stable complex between the inhibitor and the enzyme, and generation of a small C-terminal fragment of the inhibitor due to cleavage at the reactive site by tissue kallikrein.Specifications
P29622 | |
0.12 ng/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
5267 | |
<12% | |
Pre-coated 96 well plates, Adhesive Plate Covers, Assay Diluent Concentrate, Biotinylated Detection Antibody, SAV-HRP, Stop Solution, Standard, Wash Buffer, Chromogen | |
5 x 96 Tests | |
Other Research | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
0.143 ng/mL to 35 ng/mL | |
0.12 ng/mL to 35 ng/mL | |
Biotin | |
Proteases, Peptidases & Inhibitors | |
This ELISA pair antibody detects human Serpin A4. Other species not determined. | |
Human | |
Colorimetric Microplate Reader | |
KAL,KLST,KST,PI-4,PI4,kallistatin | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L | |
KAL, KLST, KST, PI-4, PI4, kallistatin | |
1 hr. 20 min. |