missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Septin-10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA563284
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MASSEVARHL LFQSHMATKT TCMSSQGSDD EQIKRENIRS LTMSGHVG Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 31%.
RAPGEF6 is a guanine nucleotide exchange factor (GEF) for Rap1A, Rap2A and M-Ras GTPases. RAPGEF6 does not interact with cAMP.Specifications
Septin-10 | |
Polyclonal | |
Unconjugated | |
SEPT10 | |
4921515A04Rik; 9430099J10Rik; AA408298; AI874685; FLJ11619; SEPT10; septin 10; SEPTIN10; septin-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P0V9 | |
SEPT10 | |
Recombinant protein corresponding to Human Septin-10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only