missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559990
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGRHQHGSHG GLDIVIIEQE DDSDRHLAQH LNNDRNPLFT Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 31%.
The protein encoded by this gene is the predominant protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.Specifications
SEMG1 | |
Polyclonal | |
Unconjugated | |
SEMG1 | |
Alpha-inhibin-31; Alpha-inhibin-92; BB115391; Cancer/testis antigen 103; CT103; dJ172H20.2; MGC14719; RATSVPIIA; semen coagulating protein; semenoclotin; semenogelin I; Semenogelin-1; SEMG; SEMG1; Seminal basic protein; seminal vesicle protein, secretion 1; seminal vesicle protein, secretion 2; seminal vesicle secretory protein 2; seminal vesicle secretory protein II; SGI; SgI-29; Svp1; Svp-1; Svp5; SVPIIA; SVS II; Svs2; SVS2P; Svs2p2; Svs5; SvsII | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6406 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P04279 | |
SEMG1 | |
Recombinant protein corresponding to Human SEMG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only