Learn More
Thermo Scientific™ Screw Vial Caps and Seals
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Brand: Thermo Scientific™ 60180-719
Code : 16
Additional Details : Weight : 1.00000kg
Description
Thermo Scientific™ Screw Vial Caps and Seals are offered in a range of colors, with and without assembled septa, for use with a broad range of autosamplers.
Specifications
White/Red | |
Black | |
Red PTFE/white silicone seal |
Black cap with red PTFE/white silicone seal | |
PTFE, Silicone | |
Thermo Scientific Spectra System AS3500/AS3000/AS1000/Surveyor/Accela LC, Agilent 1090, PE Autosystem |