missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Reusable Sterility Testing System: Silicone Gasket
Silicone Gasket for use with Reusable Sterility Testing System.
£7.35 - £8.36
Specifications
Disposable | Reusable |
---|---|
Material | Silicone |
Sterility | Non-sterile |
Type | Gasket |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10603582
|
Sartorius
6980124 |
Combisart System |
£8.36
Each |
In Stock
Log in to see stock. |
|||||
10253191
|
Sartorius
6980573 |
Reusable Sterility Testing System |
£7.35
Each |
In Stock
Log in to see stock. |
|||||
Description
Silicone Gasket - 14-555-924- Flat
- Placed under frit
- 36/47mm
Specifications
Reusable | |
Silicone | |
Non-sterile | |
Gasket |