Learn More
Sartorius Microsart™ Funnel Dispenser
Funnel dispenser for secure removal of single, sterile Microsart @filter
Brand: Sartorius 16A08
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Funnel dispenser for secure removal of single, sterile Microsart @filter
Microsart @filter units are available as packaged on trays for the use in clean benches. The packaging in bags is specially developed for the use with the Microsart Funnel Dispenser. The Funnel Dispenser for secure removal of single, sterile Microsart filter units has proven itself in practice. Even after opening the bag, the remaining funnels are protected from secondary contamination.
- Made of high-grade stainless steel
- Dispenser opening is made of polypropylene and contains a silicone O-ring
- All these materials guarantee reliable autoclaving
Specifications
Funnel Dispenser | |
High-Grade Stainless Steel | |
Funnel Dispenser |