missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kimberly-Clark™ Kimtech™ Satin Plus Latex Ambidextrous Gloves
Kimtech™ Satin Plus Latex Ambidextrous Gloves feature a smooth finish that is ideal for double donning and delivers sensitivity and performance in many applications. The non-sterile latex gloves are also rigorously tested and feature a beaded cuff.
Specifications
Certifications/Compliance | PPE Category III, EN420:2003 EN374-1 Chemical Splash and EN374-2 Microbiological Hazards ISO 9001 and ISO 13485 |
---|---|
Classification | PPE Category III |
Color | Natural |
Disposable | Single-use |
Exterior Finish | Smooth |
Description
- Assured Compliance:
- PPE Cat III according to Regulation (EU) 2016/425
- EN ISO 374-1:2016 Type C (K) Chemical Splash protection
- EN 374-4:2003 Resistance to degradation by chemicals
- EN ISO 374-5:2016 Micro Organism and Virus Protection
- Superb quality Satin Plus latex material provides high levels of contamination protection against micro-organisms, chemical splash, viruses and more.
- Suitable for use in a range of laboratories, and provided with supporting technical information to demonstrate regulatory compliance.
- Gloves are non-sterile, ambidextrous, disposable and white in color.
- The smooth finish provides added comfort, excellent tactile sensitivity for precision applications, and makes the gloves suitable for double donning.
- Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
- Made from natural rubber latex for a low dermatitis risk and containing no powder, reducing the risks of skin irritation for the wearer.
Specifications
PPE Category III, EN420:2003 EN374-1 Chemical Splash and EN374-2 Microbiological Hazards ISO 9001 and ISO 13485 | |
Natural | |
Smooth | |
Yes | |
Latex | |
Powder Free | |
Beaded Cuff | |
For use across a wide variety of non-sterile applications |
PPE Category III | |
Single-use | |
Ambidextrous | |
No | |
0.13 mm | |
Examination Gloves | |
Textured | |
Beaded/Rolled |