missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals

Product Code. 11332287
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25779-13-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00066256
InChI Key PWMWNFMRSKOCEY-MRVPVSSYSA-N
Synonym s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
PubChem CID 643312
IUPAC Name (1S)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 64°C to 66°C
Boiling Point 272°C to 274°C
Quantity 1 g
Beilstein 3196197
Optical Rotation +39° (c=3 in Ethanol)
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material (S)-(+)-Phenyl-1,2-ethanediol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'