missing translation for 'onlineSavingsMsg'
Learn More

(+)-Rutin trihydrate, 95%, Thermo Scientific Chemicals

Catalog Number p-7021292
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
250249-75-3
C27H36O19
664.566
MFCD00149490
NLLBWFFSGHKUSY-JPRRWYCFSA-N
rutoside trihydrate, +-rutin trihydrate, rutin trihydrate, rutin hydrate, unii-rf4n03853g, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-3-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyltetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy-4h-chromen-4-one trihydrate, rutoside trihydrate ep, rutin hydrate jan, rutin trihydrate hplc, rutin trihydrate, analytical standard
16218542
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)O)C5=CC(=C(C=C5)O)O)O)O)O)O)O)O.O.O.O
This item is not returnable. View return policy
250249-75-3
C27H36O19
664.566
MFCD00149490
NLLBWFFSGHKUSY-JPRRWYCFSA-N
rutoside trihydrate, +-rutin trihydrate, rutin trihydrate, rutin hydrate, unii-rf4n03853g, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-3-2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyltetrahydro-2h-pyran-2-yl oxy methyl tetrahydro-2h-pyran-2-yl oxy-4h-chromen-4-one trihydrate, rutoside trihydrate ep, rutin hydrate jan, rutin trihydrate hplc, rutin trihydrate, analytical standard
16218542
2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one;trihydrate
CC1C(C(C(C(O1)OCC2C(C(C(C(O2)OC3=C(OC4=CC(=CC(=C4C3=O)O)O)C5=CC(=C(C=C5)O)O)O)O)O)O)O)O.O.O.O

Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Rutin is the major mutagenic component of red wine, It is used in the activation of a mutagen by treatment both liver microsomal enzymes and fecal enzymes.

Solubility
Soluble in DMSO, acetone, dichloromethane or ethyl acetate. Insoluble in water.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 184°C (decomposition)
Quantity 25 g
Beilstein 75455
Sensitivity Light sensitive
Merck Index 14,3802
Solubility Information Soluble in DMSO,acetone,dichloromethane or ethyl acetate. Insoluble in water.
Optical Rotation +9° (c=0.5 in Ethanol)
Formula Weight 664.58 (610.52 Anhydrous)
Percent Purity 97%
Chemical Name or Material (+)-Rutin trihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'