Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand: SVL™ RF64
Code : NEW
Additional Details : Weight : 0.81000kg
Description
- All glass and PTFE pathway is highly resistant to chemical attack
- Can be used at temperatures up to 200°C
- System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
- Joint can be retained together with spring loaded metal clip
- Component parts RM, RF, RO and RC should be ordered separately
Specifications
Female joint | |
Glass, PTFE | |
5/Pk. | |
64 |