missing translation for 'onlineSavingsMsg'
Learn More

RNF34 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10629-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16877123

  • £130.00 / 20µL
Estimated Shipment: 01-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene contains a RINF finger, a motif known to be involved in protein-protein and protein-DNA interactions. This protein interacts with DNAJA3/hTid-1, which is a DnaJ protein reported to function as a modulator of apoptosis. Overexpression of this gene in Hela cells was shown to confer the resistance to TNF-alpha induced apoptosis, suggesting an anti-apoptotic function of this protein. This protein can be cleaved by caspase-3 during the induction of apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

RNF34
Polyclonal
Unconjugated
RNF34
CARP 1, Caspase regulator CARP1, FYVE RING finger protein Momo, hRFI, RFI, RIF, RIFF, ring finger protein 34, RING finger protein RIFF, RNF34
Rabbit
Antigen Affinity Chromatography
RUO
282845, 80196, 80751
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q6AYH3, Q969K3, Q99KR6
Rnf34
RNF34 Fusion Protein Ag0982
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers