Learn More
Ansell Ringers R074 PVC-Coated Chemical Resistant/Waterproof Impact Gloves
Developed with Ringers' F3 Technology™ and patented TPR design for increased dexterity and comfort. Textured PVC coating on the palm enhances grip capabilities, while also providing chemical resistance.
£655.00
Specifications
ANSI Abrasion Rating ANSI 5
Certifications/Compliance (Europe) EN 374_2003 Microorganism, EN 420:2003 + A1:2009
Coating Material PVC
Color Hi-vis Red
Cut Resistance Rating ANSI A3
View More Specs
Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
16679063
View Documents
Ansell
074-08
Small
£655.00
12 pairs
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
16689063
View Documents
Ansell
074-10
Large
£655.00
12 pairs
Estimated Shipment: 19-05-2025
to see stock.
Add to basket
16699063
View Documents
Ansell
074-12
2X-Large
£655.00
12 pairs
Estimated Shipment: 03-06-2025
to see stock.
Add to basket
Description
Description
  • TPR impact protection on top of the hand and over the full length of the fingers
  • PVC coating is waterproof and provides resistance to chemicals
  • Inner liner provides cut resistance throughout the entire glove
  • Durable textured palm coating enhances grip capabilities
  • Short gauntlet-style cuff keeps debris away from the hands
  • Level 1 Contact Heat performance rating at an outside exposure of 100°C, in accordance with test method EN 407
Specifications
Specifications
ANSI 5
PVC
ANSI A3
DuPont™ Aramid Fibers
Type A
Yes
R-074
EN 374_2003 Microorganism, EN 420:2003 + A1:2009
Hi-vis Red
Fully Textured
4X41CP
AJKLMO
Gauntlet
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Ansell Ringers R074 PVC-Coated Chemical Resistant/Waterproof Impact Gloves