Learn More
Epredia™ Cytoseal™ Mountant; 60, 280 and XYL
Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ 60, 280 and XYL mounting media versions.
Brand: Epredia™ 8312-4
Code : A0
Additional Details : CAS Number : 1330-20-7 Weight : 1.47600kg Transport : UN number : 1866 Chem class : 3 Pack group : II
Description
- Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles
- Cytoseal 280 mountant is a high-viscosity medium for applications where minimal spreading is desired or applications without a cover glass
- Cytoseal XYL mountant is a xylene-based rapid drying medium that allows for microscopic examination of slides soon after application
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
Specifications
4 oz. | |
Mountant |
Safety and Handling
- Cytoseal XYL
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Reproductive toxicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H315-Causes skin irritation.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P201-Obtain special instructions before use.