missing translation for 'onlineSavingsMsg'
Learn More

REXO2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen REXO2
Concentration 0.13 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16814844
View Documents Promotion Details
Proteintech
15598-1-AP-20UL
20 μL
£135.00
20µL
Please to purchase this item. Need a web account? Register with us today!
16804844
View Documents Promotion Details
Proteintech
15598-1-AP-150UL
150 μL
£349.00
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Rex2 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.
Specifications

Specifications

REXO2
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
Q9Y3B8
100043034, 100362123, 25996
REXO2 Fusion Protein Ag7976
Primary
-20°C
REXO2
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CGI 114, REX2, REXO2, RFN, RNA exonuclease 2 homolog, SFN, Small fragment nuclease, SMFN
Rex2, REXO2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications