Learn More
Restek™ Septum Nuts for use with Agilent GCs
For use with Agilent 5890, 6850, 6890, 7890 split/splitless injectors.
£60.50 - £65.34
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12902569
|
Restek™
20631 |
Manual Injection (for 26-Gauge Needles) |
£60.50
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
13187022
|
Restek™
21309 |
Autosampler and PTV (for 23-Gauge Needles) |
£65.34
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
Description
- Needle guide allows easy penetration and prevents septum coring
- Made of high-quality stainless steel
- Manual injection septum nut allows use of 26-gauge needles for on-column injections
- Autosampler and PTV septum nut allows use of 23-gauge needles
Specifications
Septum Nut |