missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Replacement SilTite™ Mini Union Ferrules
SilTite Mini Union delivers an easy, leak-free installation for capillary columns without the use of any tools
£73.00
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
12947111
|
Trajan Scientific and Medical™
073471 |
0.32 mm |
£73.00
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
12957111
|
Trajan Scientific and Medical™
073472 |
0.45 to 0.53 mm |
£73.00
Pack of 10 |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
Description
- Specifically designed for connecting capillary columns
- Connects guard columns to capillary columns
- Inert and precise glass lined bore
- Low dead volume
Specifications
Mini Union Ferrules | |
10/Pk. |