Learn More
Restek™ Replacement Chemical Traps for Agilent™ Gas Chromatographs
Easy to install replacement chemical traps attach to the same fittings as original Agilent GC equipment
Brand: Restek™ 22820
Additional Details : Weight : 0.50000kg
Description
- Built-in frits and adsorbents remove both moisture and hydrocarbons
- Trap can be regenerated to remove contaminants and restore performance
- Also available is a Replacement 32 in. Split Line with all installation hardware included
Specifications
Replacement Chemical Trap | |
Gas Chromatography | |
Agilent 6890/6850 GCs (replacement for G1544-80550) |