missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Gram Safranin
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
Brand: Thermo Scientific™ R40058
44.42 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Gram Safranin is a counterstain used in Gram stain procedures.
- For qualitative procedures to differentiate gram-positive from gram-negative microorganisms
- Selected Gram stains are CE marked
Specifications
Safranin | |
250 mL | |
Each | |
CE Marked |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.