missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Monkey GM CSF Protein
A cDNA sequence encoding the GM CSF was constructed and used to recombinantly synthesize the protein.
£99.15 - £3220.00
Specifications
Name | Monkey GM CSF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 10,000,000 IU/mg. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15914518
|
enQuireBio™
QP10647-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994508
|
enQuireBio™
QP10647-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15904518
|
enQuireBio™
QP10647-1MG |
1 mg |
£3220.00
1mg |
Estimated Shipment: 02-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Monkey GM CSF Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
APARSPSPGT QPWEHVNAIQ EARRLLNLSR DTAAEMNKTV EVVSEMFDLQ EPSCLQTRLE LYKQGLQGSL TKLKGPLTMM ASHYKQHCPP TPETSCATQI ITFQSFKENL KDFLLVIPFD CWEPVQE | |
Greater than 98.0% as determined by SDS-PAGE and RP-HPLC. |
Research Use Only | |
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 0.1ng/ml, corresponding to a specific activity of > 10,000,000 IU/mg. | |
Donkey | |
Untagged | |
GM-CSF was lyophilized from a concentrated (1 mg/ml) solution containing 1x PBS pH 7.4. |