missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Recal Color Coded Buffer Solution pH 10 at 25 °C, Reagecon
Guaranteed stability throughout entire shelf life even after opening the bottle, NIST traceable
Brand: Reagecon 10C65
Code : NEW
Additional Details : Weight : 0.54000kg
Description
- Tested by ISO 17025 accredited test method
Specifications
pH buffer standard | |
10 ±0.01 | |
6 x 90 mL | |
Yes |
Blue | |
Liquid | |
HDPE bottle |