missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Rat (Wistar) Cryopreserved Hepatocytes, Suspension Male

Rat (Wistar) Cryopreserved Hepatocytes, Suspension Male

Brand:  Gibco™ WICS10

Product Code. 13385664

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

These cryopreserved male rat (Wistar) hepatocytes have been characterized for general Phase I and Phase II drug metabolizing enzyme activities.

Cells per vial: 4–8 million

Get the Results You Need with GIBCO™ Hepatocytes
High quality hepatocytes with high viabilities, in vivo-like enzyme expression levels, and proper cell morphology increase the ability to draw in vitroin vivo correlations and make sound decisions regarding a compound's fate. Experienced technicians and proprietary isolation techniques combined with stringent release specifications ensure that you receive the highest quality cryopreserved hepatocytes.

Each lot is tested for:

  • Phase I and II enzyme activities
  • Proper morphology
  • Viabilities generally >75%

    For research use only. Not intended for human or animal therapeutic or diagnostic use.

    Related Links:

    Learn more about our cryopreserved hepatocytes.

    Learn about our full line of hepatocytes.

    Note:
    These products require shipment in LN2 vapor dewars therefore additional fees may apply.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    135°C
    Cryopreserved
    Hepatocytes
    Suspension Male Rat (Wistar) Cryopreserved Hepatocytes, 1mL, Rat Species, Hepatocytes Cell Type, Pooled Donor, Liquid Nitrogen Storage, For Hepatocytes and Hepatic Models, Pharma and Biopharma, Primary Cell Culture, Target-Based ADME/Tox Assays
    Hepatocytes
    Cell culture, drug discovery and development, target-based ADME/Tox assays, cellular toxicology assays, primary cell culture, hepatocyte culture, hepatocytes and hepatic models
    1mL
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.