missing translation for 'onlineSavingsMsg'
Learn More

RAPSN Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA558801

Product Code. 15766671

  • £431.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Immunogen sequence: MECCEESMKI ALQHGDRPLQ ALCLLCFADI HRSRGDLETA FPRYDSAMSI MTEIGNRLGQ VQALLGVAKC WVARKALDKA LDAIE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

This gene encodes a member of a family of proteins that are receptor associated proteins of the synapse. The encoded protein contains a conserved cAMP-dependent protein kinase phosphorylation site, and plays a critical role in clustering and anchoring nicotinic acetylcholine receptors at synaptic sites by linking the receptors to the underlying postsynaptic cytoskeleton, possibly by direct association with actin or spectrin. Mutations in this gene may play a role in postsynaptic congenital myasthenic syndromes. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RAPSN
Polyclonal
Unconjugated
RAPSN
43 kD receptor-associated protein of the synapse; 43 kda postsynaptic protein; 43 kDa receptor-associated protein of the synapse; 43kDa; 43kDa acetylcholine receptor-associated protein; acetylcholine receptor-associated 43 kda protein; AChR associated protein at synapses; acetylcholine receptor associated protein at synapses; 43kDa protein; likely to cluster nicotinic AChRs; CMS11; CMS1D; CMS1E; CMS4C; FADS; MGC3597; Nraps; Raps; Rapsn; rapsyn; receptor associated protein of the synapse; receptor-associated protein of the synapse; receptor-associated protein of the synapse, 43 kDa; receptor-associated protein of the synapse, 43kD; RING finger protein 205; RNF205; twitch once; two
Rabbit
Antigen affinity chromatography
RUO
5913
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry
0.08 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q13702
RAPSN
Recombinant protein corresponding to Human RAPSN
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only