missing translation for 'onlineSavingsMsg'
Learn More

Quickfit™ Borosilicate Glass Vacuum Filter Flask with Screw Thread Connector And Ground Socket

Complies with ISO 6556

Brand:  Quickfit FB1L/4SC

Code : NEW

Additional Details : Weight : 1.63000kg

Product Code. 15557810

  • £825.00 / Pack of 10
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Complies with ISO 6556
  • Büchner flasks with precision-ground Quickfit™ socket
  • Side-arm features a PTFE screwthread connector for the safe attachment of 9mm bore vacuum tubing; Unscrew connector from flask and fit tubing, rescrew connector and tubing to flask
  • Heavy walls and bases for vacuum filtration work
Specifications

Specifications

ISO 6556
Borosilicate Glass
PTFE screw thread connector
1000 mL
227 mm
136 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers