Learn More
Quickfit™ Borosilicate Glass Still Head Adapter
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
Brand: Quickfit SH4/55
Code : NEW
Detalles adicionales : Weight : 0.12000kg
Descripción
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
- Used in distillation assemblies for connecting flasks to condenser
- Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
Especificaciones
Connecting flask and condenser | |
Still Head Adapter | |
Borosilicate Glass | |
Cone size: 34/35 (flask), 34/35 (condenser); Socket Size: 14/23 |