missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrene, 98%, Thermo Scientific Chemicals
£33.50 - £303.00
Chemical Identifiers
CAS | 129-00-0 |
---|---|
Molecular Formula | C16H10 |
Molecular Weight (g/mol) | 202.25 |
MDL Number | MFCD00004136 |
InChI Key | BBEAQIROQSPTKN-UHFFFAOYSA-N |
Synonym | benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene |
PubChem CID | 31423 |
ChEBI | CHEBI:39106 |
IUPAC Name | pyrene |
SMILES | C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10673132
|
Thermo Scientific
180830250 |
25 g | Glass bottle |
£33.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245101
|
Thermo Scientific Acros
180831000 |
100 g | Glass bottle |
£84.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465755
|
Thermo Scientific Acros
180835000 |
500 g | Glass bottle |
£303.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
129-00-0 | |
202.25 | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
31423 | |
pyrene |
C16H10 | |
MFCD00004136 | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
CHEBI:39106 | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 |
Specifications
129-00-0 | |
Green-Yellow to Yellow | |
210°C | |
97.5% min. (GC) | |
MFCD00004136 | |
04,414 | |
benzo def phenanthrene, pyren, beta-pyrene, .beta.-pyrene, pyren german, unii-9e0t7wfw93, ccris 1256, pyrene def phenanthrene, coal tar pitch volatiles:pyrene | |
BBEAQIROQSPTKN-UHFFFAOYSA-N | |
pyrene | |
31423 | |
202.25 | |
Crystals and/or Chunks |
148°C to 152°C | |
393°C | |
Authentic | |
C16H10 | |
05,693 | |
15,8074 | |
Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,ether,benzene and toluene,slightly soluble in carbon tetrachloride | |
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2 | |
202.25 | |
CHEBI:39106 | |
98% | |
Pyrene |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-927-3
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Pyrene
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
RUO – Research Use Only